Anti-IFNAR1 Rabbit Monoclonal Antibody (PE (Phycoerythrin)) [clone: 110]

Supplier: Sino Biological

50469-R110-P-25 50469-R110-P-100
103630-082EA 96.92 USD
103630-082 103632-262
Anti-IFNAR1 Rabbit Monoclonal Antibody (PE (Phycoerythrin)) [clone: 110]
Antibodies
Anti-IFNAR1 Antibody (PE)

Type: Primary
Antigen: IFNAR1
Clonality: Monoclonal
Clone: Clone: 110
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR