Anti-CD55 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50468-T16-50 50468-T16-100 50468-T16-200
103589-134EA 106.62 USD
103589-134 103594-594 103595-012
Anti-CD55 Rabbit Polyclonal Antibody
Antibodies
Anti-CD55 / DAF Antibody

Type: Primary
Antigen: CD55
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR