Anti-CD55 Rabbit Monoclonal Antibody [clone:076] FITC (Fluorescein Isothiocyanate)

Supplier: Sino Biological
50468-R076-F-25 50468-R076-F-100
103630-076EA 70.7 USD
103630-076 103632-256
Anti-CD55 Rabbit Monoclonal Antibody [clone:076] FITC (Fluorescein Isothiocyanate)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD55 / DAF (rM CD55 / DAF; Catalog#50468-M08H; Q61475; Met1-Thr361) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.

Type: Primary
Antigen: CD55
Clonality: Monoclonal
Clone: Clone: 076
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR