Anti-CD55 Rabbit Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 076]

Supplier: Sino Biological

50468-R076-C-100 50468-R076-C-25
103632-254EA 258.46 USD
103632-254 103629-538
Anti-CD55 Rabbit Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 076]
Antibodies
Anti-CD55 / DAF Antibody (PerC

Type: Primary
Antigen: CD55
Clonality: Monoclonal
Clone: Clone: 076
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR