Anti-CD55 Rabbit Monoclonal Antibody [clone:076] APC (Allophycocyanin)

Supplier: Sino Biological
50468-R076-A-100 50468-R076-A-25
103629-536EA 90.9 USD
103629-536 103632-252
Anti-CD55 Rabbit Monoclonal Antibody [clone:076] APC (Allophycocyanin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD55 / DAF (rM CD55 / DAF; Catalog#50468-M08H; Q61475; Met1-Thr361) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: CD55
Clonality: Monoclonal
Clone: Clone: 076
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR