Anti-JAM-C Rabbit Polyclonal Antibody

Supplier: Sino Biological

50465-RP02-50 50465-RP02-100 50465-RP02-200
103594-588EA 106.62 USD
103594-588 103658-104 103658-106
Anti-JAM-C Rabbit Polyclonal Antibody
Antibodies
Anti-JAM-C Antibody

Type: Primary
Antigen: JAM-C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR