Anti-ICAM1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50440-RP01-100 50440-RP01-400 50440-RP01-200
103660-626EA 562.97 USD
103660-626 103658-064 103658-062
Anti-ICAM1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse ICAM1 / CD54 (rM ICAM-1; Catalog#50440-M08H; NP_034623.1; Met 1-Asn 485). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: ICAM-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR