Anti-SerpinA11 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50435-RP02-200 50435-RP02-50 50435-RP02-100
103594-958EA 142.96 USD
103594-958 103658-054 103658-052
Anti-SerpinA11 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse SerpinA11 (rM SerpinA11; Catalog#50435-M08H; NP_955018.2; Met 1-Gly 424). SerpinA11 specific IgG was purified by mouse SerpinA11 affinity chromatography.

Type: Primary
Antigen: SERPINA11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR