Anti-SerpinA11 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50435-RP01-100 50435-RP01-400 50435-RP01-200
103658-046EA 270.2 USD
103658-046 103658-048 103658-050
Anti-SerpinA11 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse SerpinA11 (rM Serpin A11; Catalog#50435-M08H; NP_955018.2; Met 1-Gly 424). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: SERPINA11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR