Anti-CHEK2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50432-RP02-200 50432-RP02-50 50432-RP02-100
103594-954EA 99 USD
103594-954 103658-042 103658-040
Anti-CHEK2 Rabbit Polyclonal Antibody
Antibodies
Anti-CHEK2 Antibody

Type: Primary
Antigen: CHEK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR