Anti-CD48 Rabbit Monoclonal Antibody [clone:405]

Supplier: Sino Biological
50415-R405-100 50415-R405-1 50415-R405-50
103594-916EA 150.18 USD
103594-916 103658-004 103589-106
Anti-CD48 Rabbit Monoclonal Antibody [clone:405]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD48 / Blast-1 (rM CD48 / Blast-1; Catalog#50415-M08H; NP_031675.1; Met1-Arg216).

Type: Primary
Antigen: Anti-CD48 Antibody
Clonality: Monoclonal
Clone: 405
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR