Anti-F3 Rabbit Monoclonal Antibody [clone:034]

Supplier: Sino Biological
50413-R034-100 50413-R034-50
103594-910EA 150.18 USD
103594-910 103657-990
Anti-F3 Rabbit Monoclonal Antibody [clone:034]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Coagulation Factor III / Tissue Factor / CD142 / F3 (rM Coagulation Factor III / Tissue Factor / CD142 / F3; Catalog#50413-M08H; NP_034301.3; Met 1-Glu 251).

Type: Primary
Antigen: CD142
Clonality: Monoclonal
Clone: 034
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR