Anti-PECAM1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50408-T16-50 50408-T16-200 50408-T16-100
103589-100EA 142.96 USD
103589-100 103594-896 103594-894
Anti-PECAM1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CD31 / PECAM-1 (Catalog#50408-M08H; NP_033868.3; Met1-Lys590). CD31 / PECAM-1 specific IgG was purified by Mouse CD31 / PECAM-1 affinity chromatography.

Type: Primary
Antigen: PECAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR