Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 4pm ET Saturday through 8pm Sunday.

Anti-PECAM1 Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 008]

Supplier: Sino Biological

50408-R008-F-25 50408-R008-F-100
103630-060EA 53.85 USD InStock
103630-060 103632-232
Anti-PECAM1 Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 008]
Antibodies
Anti-CD31 Antibody (FITC)

Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: Clone: 008
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty