Anti-ENG Rabbit Monoclonal Antibody [clone:001] APC (Allophycocyanin)

Supplier: Sino Biological
50407-R001-A-100 50407-R001-A-25
103632-224EA 151.5 USD
103632-224 103630-052
Anti-ENG Rabbit Monoclonal Antibody [clone:001] APC (Allophycocyanin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Endoglin / CD105 / ENG (rM Endoglin / CD105 / ENG; Catalog#50407-M08H; NP_031958.2; Met1-Gly580) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: ENG
Clonality: Monoclonal
Clone: Clone: 001
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR