Anti-GRN Rabbit Monoclonal Antibody [clone:034]

Supplier: Sino Biological
50396-R034-1 50396-R034-100 50396-R034-50
103657-934EA 271.4 USD
103657-934 103589-090 103594-860
Anti-GRN Rabbit Monoclonal Antibody [clone:034]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Granulin / Progranulin / GRN (rM Granulin / Progranulin / GRN; Catalog#50396-M08H; NP_032201.2; Met 1-Leu 589).

Type: Primary
Antigen: GRN
Clonality: Monoclonal
Clone: 034
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR