Anti-TIMD4 Rabbit Monoclonal Antibody [clone:102]

Supplier: Sino Biological
50388-R102-50 50388-R102-100
103594-842EA 150.18 USD
103594-842 103657-906
Anti-TIMD4 Rabbit Monoclonal Antibody [clone:102]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TIM4 / TIMD4 (rM TIM4 / TIMD4; Catalog#50388-M08H; NP_848874.3; Met 1-Thr 279).

Type: Primary
Antigen: TIM4
Clonality: Monoclonal
Clone: 102
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR