Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CPB1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50386-RP02-100 50386-RP02-200 50386-RP02-50
103657-902EA 160.46 USD InStock
103657-902 103657-904 103594-840
Anti-CPB1 Rabbit Polyclonal Antibody
Antibodies
Anti-Carboxypeptidase B1 Antib

Type: Primary
Antigen: CPB1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty