Anti-CPB1 Rabbit Monoclonal Antibody [clone:008]

Supplier: Sino Biological
50386-R008-50 50386-R008-100 50386-R008-1
103594-836EA 150.18 USD
103594-836 103659-228 103589-082
Anti-CPB1 Rabbit Monoclonal Antibody [clone:008]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carboxypeptidase B1 / CPB1 (rM Carboxypeptidase B1 / CPB1; Catalog#50386-M08H; NP_083982.1; Met1-Tyr415).

Type: Primary
Antigen: CPB1
Clonality: Monoclonal
Clone: 008
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR