Anti-AARS Rabbit Polyclonal Antibody

Supplier: Sino Biological
50384-T60-50 50384-T60-200 50384-T60-100
103594-826EA 199 USD
103594-826 103589-076 103594-824
Anti-AARS Rabbit Polyclonal Antibody
Antibodies
Anti-alanyl-tRNA synthetase An

Type: Primary
Antigen: AARS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR