Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Coagulation Factor II/F2 Protein Rabbit Polyclonal Antibody

Supplier: Sino Biological

50344-RP01-400 50344-RP01-100 50344-RP01-200
103660-596 103660-594 103660-592
Anti-Coagulation Factor II/F2 Protein Rabbit Polyclonal Antibody
Anti-Thrombin Antibody

Type: Primary
Antigen: Coagulation Factor II/F2 Protein
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty