Anti-AGT Rabbit Polyclonal Antibody

Supplier: Sino Biological
50337-RP01-400 50337-RP01-100 50337-RP01-200
103657-820EA 351.49 USD
103657-820 103657-822 103657-818
Anti-AGT Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse Angiotensinogen / SerpinA8 / AGT (Catalog#50337-M08H; P11859; Met1-Val477). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: AGT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR