Anti-ECM1 Rabbit Monoclonal Antibody [clone:001]

Supplier: Sino Biological
50331-R001-100 50331-R001-50
103594-746EA 150.18 USD
103594-746 103657-792
Anti-ECM1 Rabbit Monoclonal Antibody [clone:001]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ECM1 (rM ECM1; Catalog#50331-M08H; Q61508-1; Met 1-Glu 559).

Type: Primary
Antigen: ECM1
Clonality: Monoclonal
Clone: 001
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR