Anti-PLA2G1B Rabbit Polyclonal Antibody

Supplier: Sino Biological
50329-RP01-400 50329-RP01-200 50329-RP01-100
103657-786EA 562.97 USD
103657-786 103657-782 103657-784
Anti-PLA2G1B Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse PLA2G1B (Catalog#50329-M08H; NP_035237.1; Met1-Cys146). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: PLA2G1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR