Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD16 Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 002]

Supplier: Sino Biological

50326-R002-F-25 50326-R002-F-100
103630-020 103632-196
Anti-CD16 Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 002]
Anti-CD16 Antibody (FITC)

Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: Clone: 002
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty