Anti-LDLR Rabbit Polyclonal Antibody

Supplier: Sino Biological
50305-T24-100 50305-T24-200 50305-T24-50
103594-560EA 352.03 USD
103594-560 103594-558 103589-038
Anti-LDLR Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse LDL Receptor / LDLR (rM LDL Receptor / LDLR; Catalog#50305-M08H; NP_034830.2; Met1-Arg790). LDL Receptor / LDLR specific IgG was purified by Mouse LDL Receptor / LDLR affinity chromatography.

Type: Primary
Antigen: LDLR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR