Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD25 Rabbit Monoclonal Antibody (PE (Phycoerythrin)) [clone: 051]

Supplier: Sino Biological

50292-R051-P-100 50292-R051-P-25
103632-184 103630-008
Anti-CD25 Rabbit Monoclonal Antibody (PE (Phycoerythrin)) [clone: 051]
Anti-CD25 Antibody (PE)

Type: Primary
Antigen: CD25
Clonality: Monoclonal
Clone: Clone: 051
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty