Anti-LRPAP1 Rabbit Monoclonal Antibody [clone:009]

Supplier: Sino Biological
50281-R009-100 50281-R009-50
103657-640EA 271.4 USD
103657-640 103594-506
Anti-LRPAP1 Rabbit Monoclonal Antibody [clone:009]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse LRPAP1 (rM LRPAP1; Catalog#50281-M08H; NP_038615.2; Gln 29-Leu 360).

Type: Primary
Antigen: LRPAP1
Clonality: Monoclonal
Clone: 009
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR