Anti-LTA4H Rabbit Polyclonal Antibody

Supplier: Sino Biological
50268-RP01-100 50268-RP01-400 50268-RP01-200
103657-608EA 562.97 USD
103657-608 103657-604
Anti-LTA4H Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse LTA4H / LTA4 Hydrolase (rM LTA4H; Catalog#50268-M08B; NP_032543.2; Met 1-Asp 611). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: LTA4H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR