Anti-DR6 Rabbit Polyclonal Antibody

50263-T16-50 50263-T16-200 50263-T16-100
103594-468EA 214.31 USD
103594-468 103594-466 103589-018
Anti-DR6 Rabbit Polyclonal Antibody
Anti-DR6 Antibody

Type: Primary
Antigen: DR6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR