Anti-TNFRSF21 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50263-RP01-400 50263-RP01-200 50263-RP01-100
103659-122EA 270.2 USD
103659-122 103659-124 103659-126
Anti-TNFRSF21 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse TNFRSF21 (rM TNFRSF21; Catalog#50263-M08H; NP_848704.1; Met 1-His 349). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: DR6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR