Anti-PCSK9 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50251-RP02-50 50251-RP02-100 50251-RP02-200
103657-550EA 352.03 USD
103657-550 103657-548 103594-442
Anti-PCSK9 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse PCSK9 (rM PCSK9; Catalog#50251-M08H; NP_705793.1; Met 1-Gln 694). PCSK9 specific IgG was purified by mouse PCSK9 affinity chromatography
.

Type: Primary
Antigen: PCSK9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR