Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DKK3 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50247-RP02-100 50247-RP02-200 50247-RP02-50
103657-530EA 160.46 USD InStock
103657-530 103657-532 103594-434
Anti-DKK3 Rabbit Polyclonal Antibody
Antibodies
Anti-DKK3 Antibody

Type: Primary
Antigen: DKK3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty