Anti-CST3 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50239-RP01-200 50239-RP01-400 50239-RP01-100
103657-492EA 351.49 USD
103657-492 103657-494 103657-490
Anti-CST3 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CST3 (rM CST3; Catalog#50239-M08H; NP_034106.2; Met 1-Ala 140). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CSTC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR