Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD226 Rabbit Monoclonal Antibody [clone: 26]

Supplier: Sino Biological

50232-R026-100 50232-R026-50
103657-462EA 247.69 USD InStock
103657-462 103594-404
Anti-CD226 Rabbit Monoclonal Antibody [clone: 26]
Antibodies
Anti-CD226 Antibody

Type: Primary
Antigen: CD226
Clonality: Monoclonal
Clone: Clone: 26
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty