Anti-S100A6 Rabbit Monoclonal Antibody [clone:016]

Supplier: Sino Biological
50229-R016-50 50229-R016-1 50229-R016-100
103594-396EA 150.18 USD
103594-396 103589-000 103657-444
Anti-S100A6 Rabbit Monoclonal Antibody [clone:016]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse S100A6 / Calcyclin (rM S100A6; Catalog#50229-MNAE; Met 1-Lys 89; NP_035443.1).

Type: Primary
Antigen: S100A6
Clonality: Monoclonal
Clone: 016
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR