Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-S100A13 Rabbit Monoclonal Antibody [clone: 184]

Supplier: Sino Biological

50207-R184-100 50207-R184-50
103657-374 103594-350
Anti-S100A13 Rabbit Monoclonal Antibody [clone: 184]
Anti-S100A13 Antibody

Type: Primary
Antigen: S100A13
Clonality: Monoclonal
Clone: Clone: 184
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty