Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ICOS ligand Rabbit Polyclonal Antibody

Supplier: Sino Biological

50190-RP02-200 50190-RP02-100 50190-RP02-50
103657-312 103657-310 103594-290
Anti-ICOS ligand Rabbit Polyclonal Antibody
Anti-ICOS ligand Antibody

Type: Primary
Antigen: ICOS ligand
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty