Anti-ICOSLG Rabbit Polyclonal Antibody

Supplier: Sino Biological
50190-RP01-200 50190-RP01-100
103657-306EA 270.62 USD
103657-306 103657-308
Anti-ICOSLG Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse B7H2 extracellular domain (rM B7H2; Catalog#50190-M08H; NP_056605.1; Met 1-Lys 279). Total IgG was purified by Protein A affinity chromatography
.

Type: Primary
Antigen: ICOS ligand
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR