Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FGFRL1 Rabbit Monoclonal Antibody [clone: 019]

Supplier: Sino Biological

50182-R019-50 50182-R019-100
103594-276EA 128.15 USD InStock
103594-276 103657-266
Anti-FGFRL1 Rabbit Monoclonal Antibody [clone: 019]
Antibodies
Anti-FGFRL1 Antibody

Type: Primary
Antigen: FGFRL1
Clonality: Monoclonal
Clone: Clone: 019
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty