Anti-ACVR2B Rabbit Polyclonal Antibody

Supplier: Sino Biological
50173-RP01-200 50173-RP01-100 50173-RP01-400
103657-248EA 229.88 USD
103657-248 103657-250 103657-252
Anti-ACVR2B Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse ACVR2B extracellular domain (rM ACVR2B; Catalog#50173-M08H; NP_031423.1; Met 1-Thr 134). Total IgG was purified by Protein A affinity chromatography
.

Type: Primary
Antigen: ACVR2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR