Anti-GFRA1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50171-RP02-100 50171-RP02-50 50171-RP02-200
103594-264EA 142.96 USD
103594-264 103657-240 103657-242
Anti-GFRA1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse GFRa1 / GFR alpha-1 (rM GFRA1; Catalog#50171-M08H; AAB86600.1; Met 1-Ser 425). GFRa1 specific IgG was purified by mouse GFRa1 affinity chromatography
.

Type: Primary
Antigen: GFRA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR