Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RBP4 Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 008]

Supplier: Sino Biological

50170-R008-F-100 50170-R008-F-25
103632-148 103631-420
Anti-RBP4 Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 008]
Anti-RBP4 Antibody (FITC)

Type: Primary
Antigen: RBP4
Clonality: Monoclonal
Clone: Clone: 008
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty