Anti-VCAM1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50163-RP01-100 50163-RP01-400 50163-RP01-200
103657-196EA 351.49 USD
103657-196 103657-198 103657-194
Anti-VCAM1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse VCAM-1 / CD106 (rM VCAM-1 / CD106; Catalog#50163-M08H; NP_035823.3; Met 1-Glu 698). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: VCAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR