Anti-TNFRSF19 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50148-RP02-200 50148-RP02-100 50148-RP02-50
103657-156EA 352.03 USD
103657-156 103657-154 103594-214
Anti-TNFRSF19 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse TNFRSF19 (rM TNFRSF19; Catalog#50148-M08H; NP_001157627.1; Met 1-Leu 170). TNFRSF19 specific IgG was purified by mouse TNFRSF19 affinity chromatography
.

Type: Primary
Antigen: TNFRSF19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR