Anti-TNFRSF19 Rabbit Monoclonal Antibody [clone:104]

Supplier: Sino Biological
50148-R104-50 50148-R104-100 50148-R104-1
103657-144EA 271.4 USD
103657-144 103588-950 103594-210
Anti-TNFRSF19 Rabbit Monoclonal Antibody [clone:104]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TNFRSF19 / TROY (rM TNFRSF19 / TROY; Catalog#50148-M08H; NP_001157627.1; Met1-Leu170).

Type: Primary
Antigen: TNFRSF19
Clonality: Monoclonal
Clone: 104
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR