Anti-TFPI Rabbit Monoclonal Antibody [clone:107]

Supplier: Sino Biological
50131-R107-50 50131-R107-100
103657-064EA 271.4 USD
103657-064 103594-156
Anti-TFPI Rabbit Monoclonal Antibody [clone:107]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TFPI (rM TFPI; Catalog#50131-M08H; NP_035706.1; Met 1-Lys 289).

Type: Primary
Antigen: TFPI
Clonality: Monoclonal
Clone: 107
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR