Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Serum Amyloid P Rabbit Polyclonal Antibody

Supplier: Sino Biological

50113-RP01-100 50113-RP01-200 50113-RP01-400
103656-970 103656-968 103656-972
Anti-Serum Amyloid P Rabbit Polyclonal Antibody
Anti-Serum Amyloid P Antibody

Type: Primary
Antigen: Serum Amyloid P
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty