Anti-CD27 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50110-RP01-100 50110-RP01-200 50110-RP01-400
103656-954EA 351.49 USD
103656-954 103656-952 103656-956
Anti-CD27 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CD27 (rM CD27; Catalog#50110-M08H; NP_001028298.1; Met 1-Arg 182). Total IgG was purified by Protein A affinity chromatography
.

Type: Primary
Antigen: CD27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR