Anti-IL1B Rabbit Polyclonal Antibody

Supplier: Sino Biological
50101-T48-100 50101-T48-50 50101-T48-200
103594-086EA 352.03 USD
103594-086 103594-084 103588-900
Anti-IL1B Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse IL-1 beta (rM IL-1 beta; Catalog#50101-MNAE; P10749; Val118-Ser269). IL-1 beta specific IgG was purified by Mouse IL-1 beta affinity chromatography.

Type: Primary
Antigen: IL-1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR